Europium(III) perchlorate hexahydrate 50% in aqueous solution ≥99.9% (Eu basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Europium(III) perchlorate
93-6323-10G
93-6323-2G
100203-550EA
72.18
USD
100203-550
100207-302
Europium(III) perchlorate hexahydrate 50% in aqueous solution ≥99.9% (Eu basis)
Europium (III) perchlorate hexahydrate
Formula:
Cl₃EuO₁₂·6H₂O MW: 558.4 g/mol |
MDL Number:
MFCD00150233 CAS Number: 13537-22-9 UN: 3211 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...