Praseodymium(III) perchlorate 50% in aqueous solution ≥99.9% (Pr basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-5912-50G 93-5912-10G
100197-916EA 122 USD
100197-916 100209-164
Praseodymium(III) perchlorate 50% in aqueous solution ≥99.9% (Pr basis)
Praseodymium (III) perchlorate
Formula: Cl₃O₁₂Pr
MDL Number: MFCD00016286
CAS Number: 51411-03-1
UN: 3211
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR