Cerium (III) perchlorate hexahydrate, Reagent Grade

Danger

93-5860-50G 93-5860-250G
100202-230EA 70.77 USD
100202-230 100198-422
Cerium (III) perchlorate hexahydrate, Reagent Grade
Cerium (III) perchlorate hexahydrate
Formula: CeCl₃O₁₂·6H₂O
MDL Number: MFCD00150205
CAS Number: 14017-47-1
UN: 1481
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR