Cerium(III) carbonate hydrate ≥96% (Ce basis)
Supplier: STREM CHEMICALS INC MS
93-5800-1000G
93-5800-250G
93-5800-5000G
100205-894EA
455.75
USD
100205-894
100199-636
100210-714
Cerium(III) carbonate hydrate ≥96% (Ce basis)
Cerium (III) carbonate hydrate
Formula:
C₃Ce₂O₉·H₂O |
MDL Number:
MFCD00149635 CAS Number: 54451-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...