Lanthanum(III) chloride hexahydrate ≥99.9% (La basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-5731-100G
93-5731-500G
100196-772EA
111.82
USD
100196-772
100199-144
Lanthanum(III) chloride hexahydrate ≥99.9% (La basis)
Lanthanum (III) chloride hexahydrate
Formula:
Cl₃La·6H₂O Melting Pt: 91 °C (dec.) |
MDL Number:
MFCD00149755 CAS Number: 17272-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...