Hexafluoroantimonic acid 65 - 70% in aqueous solution
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Hydrogen hexafluoroantimonate(V)
93-5130-100G
93-5130-25G
100208-190EA
141.54
USD
100208-190
100208-188
Hexafluoroantimonic acid 65 - 70% in aqueous solution
Hexafluoroantimonic acid
Formula:
HF₆Sb MW: 236.758 g/mol |
MDL Number:
MFCD00011327 CAS Number: 16950-06-4 UN: 3264 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...