Cadmium perchlorate hexahydrate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-4818-25G
93-4818-100G
100210-424EA
60.86
USD
100210-424
100199-110
Cadmium perchlorate hexahydrate ≥99%
Cadmium perchlorate hexahydrate
Formula:
CdCl₂O₈·6H₂O MW: 419.4 g/mol |
MDL Number:
MFCD00150202 CAS Number: 10326-28-0 UN: 3087 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...