Potassium dicyanoargentate ≥99.9% (Ag basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Potassium silver cyanide

93-4739-50G 93-4739-10G
100198-386EA 333 USD
100198-386 100196-684
Potassium dicyanoargentate ≥99.9% (Ag basis)
Potassium dicyanoargentate
Formula: C₂AgKN₂
MDL Number: MFCD00036297
CAS Number: 506-61-6
UN: 3288
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR