Bis(cyclopentadienyl)zirconium dichloride ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Zirconocene dichloride
93-4002-25G
93-4002-100G
93-4002-5G
100203-464EA
158.52
USD
100203-464
100198-050
100210-394
Bis(cyclopentadienyl)zirconium dichloride ≥99%
Bis(cyclopentadienyl)zirconium dichloride
Metallocenes, Derivatives & Cp Precursors
Formula:
C₁₀H₁₀Cl₂Zr MW: 292.32 g/mol |
MDL Number:
MFCD00003726 CAS Number: 1291-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...