Phenylarsonic acid ≥98%

Danger

93-3322-25G 93-3322-5G
100210-370EA 209.48 USD
100210-370 100204-706
Phenylarsonic acid ≥98%
Phenylarsonic acid
Formula: C₆H₇AsO₃
MW: 202.04 g/mol
Melting Pt: 163 °C (dec.)
Density: 1.760
MDL Number: MFCD00002097
CAS Number: 98-05-5
UN: 1557
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR