Zinc (II) cyclohexanebutyrate dihydrate standard for AAS

Supplier: STREM CHEMICALS INC MS

Synonyms: Zinc cyclohexanebutyrate dihydrate

93-3033-5G
100208-152EA 101 USD
100208-152
Zinc (II) cyclohexanebutyrate dihydrate standard for AAS
Zinc (II) cyclohexanebutyrate dihydrate
Formula: C₂₀H₃₄O₄Zn·2H₂O
MDL Number: MFCD00035537
CAS Number: 38582-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR