Diethylzinc ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

93-3030-100G
100208-712EA 159 USD
100208-712
Diethylzinc ≥95%
Diethylzinc

ALD, Metal Alkyls, Volatile Precursors for CVD

Formula: C₄H₁₀Zn
Boiling Pt: 124 °C
Melting Pt: –28 °C
Density: 1,18
Flash Pt: –1 °F
MDL Number: MFCD00009021
CAS Number: 557-20-0
UN: 3394
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR