Stainless steel, type 316, powder

Supplier: STREM CHEMICALS INC MS

93-2671-500G
100200-544EA 191.08 USD
100200-544
Stainless steel, type 316, powder
Stainless steel, type 316
Formula: Fe
MW: 55.845 g/mol
Boiling Pt: 3000 °C
Melting Pt: 1535 °C
Density: 7.86
MDL Number: MFCD00148499
CAS Number: 52013-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR