Iron disilicide ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: FerrosiliconIron silicide

93-2648-50G 93-2648-10G
100199-028EA 97.66 USD
100199-028 100197-204
Iron disilicide ≥99%
Iron disilicide
Formula: FeSi₂
MW: 112.02 g/mol
Melting Pt: 1220 °C
Density: 4,75
MDL Number: MFCD00016087
CAS Number: 12022-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR