Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) ≥75% in isopropanol (IPA)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Titanium(IV) Diisopropoxide Bis(acetylacetonate)Titanium (IV) bis (acetylacetonate) diisopropoxideDiisopropoxytitanium(IV) Bis(acetylacetonate)Titanium (di-i-propoxide)bis(acetylacetonate)

93-2203-500G
100197-742EA 203.82 USD
100197-742
Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) ≥75% in isopropanol (IPA)
Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV)

Alkoxide

Formula: C₁₆H₂₈O₆Ti
MW: 364.26 g/mol
Density: 0.995
Flash Pt: 72 °F
MDL Number: MFCD00000031
CAS Number: 17927-72-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR