Calcium disilicide ≥90% (~4% Fe, ~6% Ba+Al)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Calcium silicide
93-2043-500G
93-2043-100G
93-2043-2000G
100205-404EA
665.23
USD
100205-404
100196-540
100206-956
Calcium disilicide ≥90% (~4% Fe, ~6% Ba+Al)
Calcium disilicide
Formula:
CaSi₂ MW: 96.25 g/mol |
UN:
1405 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...