Potassium wolframate ≥99.5% (W basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Potassium tungsten oxidePotassium tungstate

93-1957-50G 93-1957-10G
100207-278EA 276 USD
100207-278 100203-406
Potassium wolframate ≥99.5% (W basis)
Potassium wolframate
Formula: K₂O₄W
MW: 326.03 g/mol
MDL Number: MFCD00049668
CAS Number: 7790-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR