Potassium tripolyphosphate ≥94%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Potassium triphosphate
93-1955-1000G
93-1955-250G
100197-726EA
33.97
USD
100197-726
100201-950
Potassium tripolyphosphate ≥94%
Potassium tripolyphosphate
Formula:
K₅O₁₀P₃ MW: 448.41 g/mol Melting Pt: 620 °C Density: 2,54 |
MDL Number:
MFCD00054129 CAS Number: 13845-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...