Potassium pentaborate octahydrate ≥97%

Supplier: STREM CHEMICALS INC MS

93-1933-500G 93-1933-2000G
100204-206EA 174.24 USD
100204-206 100210-222
Potassium pentaborate octahydrate ≥97%
Potassium pentaborate octahydrate
Formula: K₂B₁₀O₁₆.8H₂O
Melting Pt: 780 °C
MDL Number: MFCD00213879
CAS Number: 12229-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR