Hexafluorophosphoric acid 60 - 70% in water

Supplier: STREM CHEMICALS INC MS
Danger

93-1531-1000G 93-1531-250G
100204-672EA 251.94 USD
100204-672 100209-022
Hexafluorophosphoric acid 60 - 70% in water
Hexafluorophosphoric acid
Formula: HF₆P
MDL Number: MFCD00011350
CAS Number: 16940-81-1
UN: 1782
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR