Bis(2-ethylhexyl)phosphate (contains some mono)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Di-(2-ethylhexyl)phosphoric acid
, Bis(2-ethylhexyl) hydrogen phosphate
93-1520-100G
93-1520-500G
100196-514EA
58.03
USD
100196-514
100199-494
Bis(2-ethylhexyl)phosphate (contains some mono)
Bis(2-ethylhexyl) phosphate
Formula:
C₁₆H₃₅O₄P MW: 322.43 g/mol Density: 0.974 |
MDL Number:
MFCD00009492 CAS Number: 298-07-7 UN: 1902 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...