Diisobutylaluminium chloride ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Di-i-butylaluminum chloride
93-1397-100G
93-1397-500G
100211-214EA
184
USD
100211-214
100208-102
Diisobutylaluminium chloride ≥97%
Diisobutylaluminium chloride
Formula:
C₈H₁₈AlCl MW: 176.663 g/mol Density: 0,905 |
MDL Number:
MFCD00008927 CAS Number: 1779-25-5 UN: 3394 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...