Aluminum acetate, basic (boric acid adduct)

Supplier: STREM CHEMICALS INC MS

93-1301-100G 93-1301-500G
100198-298EA 223.63 USD
100198-298 100210-174
Aluminum acetate, basic (boric acid adduct)
Aluminum acetate, basic (boric acid adduct)
MDL Number: MFCD00070418
CAS Number: 7360-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR