Phenylmagnesium chloride 2-3M in THF

Supplier: STREM CHEMICALS INC MS
Danger

93-1269-250MMOL 93-1269-1MOL
100196-488EA 44 USD
100196-488 100205-304
Phenylmagnesium chloride 2-3M in THF
Phenylmagnesium chloride

Grignard

Formula: C₆H₅ClMg
Flash Pt: 1 °F
MDL Number: MFCD00000464
UN: 3399
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR