Benzylmagnesium chloride 1-2 M in THF

Supplier: STREM CHEMICALS INC MS
Danger

93-1202-250MMOL 93-1202-1MOL
100200-046EA 264.68 USD
100200-046 100209-410
Benzylmagnesium chloride 1-2 M in THF
Benzylmagnesium chloride

Grignard

Formula: C₇H₇ClMg
Flash Pt: 1 °F
MDL Number: MFCD00000469
UN: 3399
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR