Disodium phthalocyanine

Supplier: STREM CHEMICALS INC MS

Synonyms: Phthalocyanine SodiumSodium PhthalocyanineSodium phthalocyaninePhthalocyanine Disodium

93-1176-5G
100206-912EA 251.94 USD
100206-912
Disodium phthalocyanine
Disodium phthalocyanine
Formula: C₃₂H₁₆N₈Na₂
MW: 558.52 g/mol
MDL Number: MFCD00066368
CAS Number: 25476-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR