Sodium borodeuteride (96% D)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1117-1G
93-1117-5G
100198-958EA
924.25
USD
100198-958
100198-956
Sodium borodeuteride (96% D)
Sodium borodeuteride
Formula:
D₄BNa MW: 41.857 g/mol Melting Pt: ~400 °C (dec.) Density: 1.074 |
UN:
1426 ADR: 4.3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...