sec-Butyllithium 12% (1.4M) in cyclohexane
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
s-Butyllithium
93-0389-1MOL
93-0389-250MMOL
100210-848EA
669.48
USD
100210-848
100205-254
sec-Butyllithium 12% (1.4M) in cyclohexane
sec-Butyllithium
Formula:
C₄H₉Li MW: 64.06 g/mol Flash Pt: 1 °F |
MDL Number:
MFCD00009323 UN: 3394 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...