Lithium hexafluorostannate(IV) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-0327-5G 93-0327-25G
100200-016EA 835.08 USD
100200-016 100210-840
Lithium hexafluorostannate(IV) ≥99%
Lithium hexafluorostannate(IV)
Formula: F₆Li₂Sn
MDL Number: MFCD00058771
CAS Number: 17029-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR