(Trimethylphosphine)gold(I) chloride ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Chlorotrimethylphosphine gold(I)
, Chlorotrimethylphosphinegold(I)
79-0850-2G
79-0850-500MG
100196-968EA
1128.06
USD
100196-968
100208-696
(Trimethylphosphine)gold(I) chloride ≥98%
(Trimethylphosphine)gold (I) chloride
Formula:
C₃H₉AuClP MW: 308.5 g/mol Melting Pt: 225…228 °C |
MDL Number:
MFCD00058861 CAS Number: 15278-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...