(Triethylphosphine)gold(I) chloride ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Chlorotriethylphosphinegold(I)

79-0750-250MG 79-0750-5G 79-0750-1G
100208-694EA 2067.88 USD
100208-694 100208-692 100208-480
(Triethylphosphine)gold(I) chloride ≥99%
(Triethylphosphine)gold (I) chloride
Formula: C₆H₁₅AuClP
MW: 350.577 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD00013205
CAS Number: 15529-90-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR