cis-Dichlorobis(triphenylphosphite)platinum(II) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: cis-Bis(triphenylphosphine)palladium (II) chloridecis-Bis(triphenylphosphine)platinum(II) chloridePt(PPh₃)₂Cl₂

78-0410-5G 78-0410-1G
100196-372EA 219.38 USD
100196-372 100196-374
cis-Dichlorobis(triphenylphosphite)platinum(II) ≥98%
cis-Dichlorobis(triphenylphosphite)platinum(II)
Formula: [(C₆H₅)₃P]₂PtCl₂
MW: 790,56 g/mol
MDL Number: MFCD00010825
CAS Number: 15604-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR