Carboplatin ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: cis-Diamine(1,1-cyclobutanedicarboxylato)platinum(II)cis-Diammine(1,1-cyclobutanedicarboxylato)platinum(II)1,1-Cyclobutanedicarboxylatodiammineplatinum(II)

78-0255-250MG
100206-812EA 246.28 USD
100206-812
Carboplatin ≥99%
Carboplatin
Formula: C₆H₁₂N₂O₄Pt
MW: 371.26 g/mol
MDL Number: MFCD00070464
CAS Number: 41575-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR