Methoxy(cyclooctadiene)iridium(I) dimer ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Bis(1,5-cyclooctadiene)di-mu-methoxydiiridium(I)
, (1,5-Cyclooctadiene)(methoxy)iridium(I) dimer
, Bis(1,5-cyclooctadiene)dimethoxydiiridium
, Di-mu-methoxobis(1,5-cyclooctadiene)diiridium(I)
77-1100-500MG
77-1100-2G
101419-118EA
1135.14
USD
101419-118
101419-116
Methoxy(cyclooctadiene)iridium(I) dimer ≥98%
Methoxy(cyclooctadiene)iridium(I) dimer
Formula:
C₁₈H₃₀Ir₂O₂ MW: 662.87 g/mol |
MDL Number:
MFCD08459360 CAS Number: 12148-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...