Tungsten boride ≥99.5% (W basis)
Supplier: STREM CHEMICALS INC MS
74-2000-10G
74-2000-50G
100198-248EA
215.14
USD
100198-248
100208-460
Tungsten boride ≥99.5% (W basis)
Tungsten boride
Formula:
BW |
MDL Number:
MFCD00049708 CAS Number: 12007-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...