Bis(cyclopentadienyl)tungsten dichloride ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
74-1200-1G
74-1200-5G
100202-420EA
670.89
USD
100202-420
100201-018
Bis(cyclopentadienyl)tungsten dichloride ≥99%
Bis(cyclopentadienyl)tungsten dichloride
Metallocenes, Derivatives & Cp Precursors
Formula:
C₁₀H₁₀Cl₂W |
MDL Number:
MFCD00269852 CAS Number: 12184-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...