Europium(III) trifluoromethanesulfonate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Europium(III) trifluoromethanesulphonate
, Europium triflate
, Europium(III) trifluoromethanesulfonate
63-1000-1G
63-1000-5G
101442-372EA
134.46
USD
101442-372
101442-370
Europium(III) trifluoromethanesulfonate ≥98%
Europium (III) trifluoromethanesulphonate
Metal Triflate
Formula:
C₃EuF₉O₉S₃ MW: 599.17 g/mol |
MDL Number:
MFCD00209587 CAS Number: 52093-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...