Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium hydrate
, [Ba(TMHD)2]
56-8500-25G
56-8500-1G
56-8500-5G
100196-958EA
619
USD
100196-958
100197-566
100207-416
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formula:
C₂₂H₃₈BaO₄·H₂O Boiling Pt: 285 °C/ 225 °C (0.05 mmHg) (dec./ subl.) Melting Pt: 195…200 °C |
UN:
1564 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...