Tin(II) acetate ≥99%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Tin(II) Acetate
50-1975-25G
50-1975-5G
100208-932EA
90.58
USD
100208-932
100200-708
Tin(II) acetate ≥99%
Tin (II) acetate
Formula:
C₄H₆O₄Sn MW: 236.8 g/mol Boiling Pt: 155 °C (0.1 mmHg) (subl.) Melting Pt: 180 °C |
MDL Number:
MFCD00054408 CAS Number: 638-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...