Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Pd(DIPHOS)2
, Pd(dppe)2
46-0200-1G
46-0200-5G
100196-262EA
137.29
USD
100196-262
100206-108
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) ≥98%
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)
Formula:
C₅₂H₄₈P₄Pd MW: 903.27 g/mol |
MDL Number:
MFCD00009880 CAS Number: 31277-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...