Allylpalladium(II) chloride dimer ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Di-mu-chlorobis[(eta-allyl)palladium(II)]Allylpalladium chloride dimerAllylpalladium(II) chloride

46-0100-2G 46-0100-500MG
100198-814EA 117.48 USD
100198-814 100206-696
Allylpalladium(II) chloride dimer ≥98%
Allylpalladium(II) chloride dimer
Formula: C₆H₁₀Cl₂Pd₂
MW: 365.89 g/mol
Melting Pt: 120 °C (dec.)
MDL Number: MFCD00044874
CAS Number: 12012-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR