Allylchloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]palladium(II) ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
46-0040-250MG
46-0040-1G
100206-694EA
352.43
USD
100206-694
100196-260
Allylchloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]palladium(II) ≥98%
Allylchloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]palladium(II)
NHC
Formula:
C₃₀H₄₁ClN₂Pd MW: 571.5330 g/mol Melting Pt: 190 °C (dec.) |
MDL Number:
MFCD08561131 CAS Number: 478980-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...