Rhodium(III) oxide, anhydrous ≥99.9% (Rh basis)
Supplier: STREM CHEMICALS INC MS
45-1865-2G
45-1865-500MG
100209-354EA
2053.72
USD
100209-354
100206-690
Rhodium(III) oxide, anhydrous ≥99.9% (Rh basis)
Rhodium (III) oxide
Formula:
O₃Rh₂ MW: 253.81 g/mol Melting Pt: 1100 °C (dec.) Density: 8.20 |
MDL Number:
MFCD00011205 CAS Number: 12036-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...