Rhodium(III) nitrate (10% Rh) in water

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Rhodium(III) nitrate

45-1808-5G
100211-110EA 1987.2 USD
100211-110
Rhodium(III) nitrate (10% Rh) in water
Rhodium (III) nitrate
Formula: N₃O₉Rh
MW: 288.92 g/mol
MDL Number: MFCD00011202
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR