Chlorobis(cyclooctene)rhodium(I) dimer ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
45-0265-1G
45-0265-250MG
100196-240EA
165.6
USD
100196-240
100201-226
Chlorobis(cyclooctene)rhodium(I) dimer ≥98%
Chlorobis(cyclooctene)rhodium(I) dimer
Formula:
C₃₂H₅₆Cl₂Rh₂ MW: 717.51 g/mol |
MDL Number:
MFCD00013287 CAS Number: 12279-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...