(-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R,R)-Me-DUPHOS-Rh

45-0160-2G 45-0160-100MG 45-0160-500MG
100208-654EA 751.57 USD
100208-654 100208-652 100197-144
(-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate ≥98%
(-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate

BPE, DUPHOS

Formula: C₂₇H₃₈F₃O₃P₂RhS
MW: 664.5021 g/mol
MDL Number: MFCD00269862
CAS Number: 187682-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR