Tris(triphenylphosphine)ruthenium(II) chloride ≥99%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Dichlorotris(triphenylphosphine)ruthenium(II)
, Dichlorotris(triphenylphosphino)ruthenium (II)
44-0500-5G
44-0500-1G
100201-644EA
332.62
USD
100201-644
100201-642
Tris(triphenylphosphine)ruthenium(II) chloride ≥99%
Tris(triphenylphosphine)ruthenium (II) chloride
Formula:
C₅₄H₄₅Cl₂P₃Ru MW: 958.85 g/mol |
MDL Number:
MFCD00013077 CAS Number: 15529-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...