[RuCl(p-cymene)((R)-dtbm-segphos®)]Cl

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Chloro{(R)-(-)-5,5'-bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole}(p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-dtbm-segphos®)]ClChloro[(R)-(-)-5,5'-bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride

44-0102-250MG
101418-866EA 75.02 USD
101418-866
[RuCl(p-cymene)((R)-dtbm-segphos®)]Cl
[RuCl(p-cymene)((R)-dtbm-segphos®)]Cl

SEGPHOS

Formula: C₈₄H₁₁₄Cl₂O₈P₂Ru
MW: 1485.75 g/mol
Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753019
CAS Number: 944451-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR