[RuCl(p-cymene)((S)-segphos®)]Cl

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Chloro[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) ChlorideChloro[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((S)-segphos®)]Cl

44-0097-250MG
101418-854EA 75.02 USD
101418-854
[RuCl(p-cymene)((S)-segphos®)]Cl
[RuCl(p-cymene)((S)-segphos®)]Cl

SEGPHOS

Formula: C₄₈H₄₂Cl₂O₄P₂Ru
MW: 916.78 g/mol
Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753017
CAS Number: 944451-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR