[RuCl(p-cymene)((R)-xylbinap)]Cl

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Chloro[(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl](p-cymene)ruthenium(II) ChlorideChloro{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}(p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-xylbinap}]Cl

44-0092-250MG
101418-834EA 90.58 USD
101418-834
[RuCl(p-cymene)((R)-xylbinap)]Cl
[RuCl(p-cymene)((R)-xylbinap)]Cl

BINAP

Formula: C₆₂H₆₂Cl₂P₂Ru
MW: 1041.10 g/mol
Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753015
CAS Number: 944451-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR